siRNA Details
Gene Symbol: 4931415C17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016458.1 | - | 1 | 509 | ||
AK088235.1 | - | 1 | 750 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716128.1 | - | 1 | 509 | Chr. 8 - 12672427 - 12677853 [+] on Build GRCm38 |
