siRNA Details
Gene Symbol: 4930421P05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077193.1 | - | 1 | 1994 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV281768.2 | - | 1 | 620 | Chr. 2 - 178408643 - 178409290 [+] on Build GRCm38 |
