siRNA Details
Gene Symbol: 1700123O21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045799.1 | 4 | 610 | |||
GenBank mRNA | AK006584.1 | - | 4 | 611 | ||
AK018984.1 | - | 4 | 612 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700123O21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045799.1 | 4 | 610 | |||
GenBank mRNA | AK006584.1 | - | 4 | 611 | ||
AK018984.1 | - | 4 | 612 |