siRNA Details
Gene Symbol: 1700123O21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045799.1 | 1 | 110 | |||
GenBank mRNA | AK006584.1 | - | 1 | 111 | ||
AK018984.1 | - | 1 | 112 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707361.1 | - | 1 | 107 | Chr. 16 - 5975590 - 5998461 [+] on Build GRCm38 |
