siRNA Details
Gene Symbol: 5730405N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017498.1 | - | 115 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717286.1 | - | 1 | 112 | Chr. 14 - 117903137 - 117904042 [+] on Build GRCm38 |
