siRNA Details
Gene Symbol: A930009E08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020797.1 | - | 1 | 706 | ||
AK020836.1 | - | 1 | 737 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM936805.1 | - | 1 | 569 | Chr. 1 - 97637530 - 97638207 [-] on Build GRCm38 |
