siRNA Details
Gene Symbol: Eapp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK145027.1 | - | 7 | 2614 | ||
AK145041.1 | - | 7 | 2614 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK039317.1 | - | 729 | Chr. - - [-] on Build |
