siRNA Details
Gene Symbol: Gm15396 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131623.1 | - | 2 | 353 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ041946.1 | - | 2 | 353 | Chr. 7 - 44580353 - 44590624 [-] on Build GRCm38 |
