siRNA Details
Gene Symbol: 2310043L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037994.1 | 3 | 648 | |||
GenBank mRNA | AK009784.1 | - | 3 | 652 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY709406.1 | - | 3 | 649 | Chr. 1 - 177641540 - 177642943 [-] on Build GRCm38 |
