siRNA Details
Gene Symbol: 1500012K07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045812.1 | 3 | 384 | |||
GenBank mRNA | AK005235.1 | - | 3 | 385 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705414.1 | - | 3 | 381 | Chr. 7 - 75308698 - 75318485 [+] on Build GRCm38 |
