siRNA Details
Gene Symbol: 2310034G01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040418.1 | 1 | 413 | |||
GenBank mRNA | AK009611.1 | - | 1 | 380 | ||
AK051784.1 | - | 1 | 400 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2310034G01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040418.1 | 1 | 413 | |||
GenBank mRNA | AK009611.1 | - | 1 | 380 | ||
AK051784.1 | - | 1 | 400 |