siRNA Details
Gene Symbol: 1110046J04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040707.1 | 5 | 619 | |||
GenBank mRNA | AK004186.1 | - | 5 | 619 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY704430.1 | - | 5 | 619 | Chr. 13 - 33936032 - 33960181 [+] on Build GRCm38 |
