siRNA Details
Gene Symbol: Rnf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139182.1 | - | 211 | |||
AK151082.1 | - | 1391 | ||||
AK153482.1 | - | 1392 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rnf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139182.1 | - | 211 | |||
AK151082.1 | - | 1391 | ||||
AK153482.1 | - | 1392 |