siRNA Details
Gene Symbol: 1700001D01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045475.1 | 4 | 449 | |||
GenBank mRNA | AK005580.1 | - | 4 | 450 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700001D01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045475.1 | 4 | 449 | |||
GenBank mRNA | AK005580.1 | - | 4 | 450 |