siRNA Details
Gene Symbol: 4930470F04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015532.1 | - | 1 | 357 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715080.1 | - | 1 | 357 | Chr. 19 - 59943740 - 59944657 [+] on Build GRCm38 |
