siRNA Details
Gene Symbol: 4930470O06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015539.1 | - | 4 | 623 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715087.1 | - | 4 | 623 | Chr. 8 - 59825649 - 59844721 [+] on Build GRCm38 |
