siRNA Details
Gene Symbol: 1700027A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038002.1 | 1 | 170 | |||
GenBank mRNA | BC048669.1 | - | 1 | 145 | ||
BC100504.1 | - | 145 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700027A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038002.1 | 1 | 170 | |||
GenBank mRNA | BC048669.1 | - | 1 | 145 | ||
BC100504.1 | - | 145 |