siRNA Details
Gene Symbol: 1700027A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038001.1 | 3 | 277 | |||
NR_038002.1 | 2 | 351 | ||||
GenBank mRNA | BC048669.1 | - | 2 | 326 | ||
BC100504.1 | - | 326 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700027A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038001.1 | 3 | 277 | |||
NR_038002.1 | 2 | 351 | ||||
GenBank mRNA | BC048669.1 | - | 2 | 326 | ||
BC100504.1 | - | 326 |