siRNA Details
Gene Symbol: Gm15704 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029019.1 | - | 316 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714100.1 | - | 3 | 312 | Chr. 6 - 145740902 - 145746270 [-] on Build GRCm38 |
