siRNA Details
Gene Symbol: Gm27198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133101.1 | - | 2, 3 | 174 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB615927.1 | - | 2, 3 | 174 | Chr. 14 - 118728650 - 118735145 [-] on Build GRCm38 |
