siRNA Details
Gene Symbol: Kalrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK086150.1 | - | 1 | 766 | ||
AK089128.1 | - | 1 | 768 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX763511.1 | - | 1 | 150 | Chr. 16 - 33974208 - 33974914 [-] on Build GRCm38 |
