siRNA Details
Gene Symbol: Gm15395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC048597.1 | - | 3 | 562 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU945707.1 | - | 3 | 565 | Chr. 8 - 117036494 - 117043621 [+] on Build GRCm38 |
