siRNA Details
Gene Symbol: 4930445K14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015392.1 | - | 598 | |||
AK142117.1 | - | 575 | ||||
BC037706.1 | - | 572 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714944.1 | - | 602 | Chr. - - [-] on Build |
