siRNA Details
Gene Symbol: Gm30790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038693.1 | - | 2 | 171 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB632473.1 | - | 2 | 171 | Chr. 16 - 76752113 - 76779092 [-] on Build GRCm38 |
