siRNA Details
Gene Symbol: 4933417C20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016840.1 | - | 1 | 278 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716515.1 | - | 1 | 278 | Chr. 1 - 160213694 - 160214505 [+] on Build GRCm38 |
