siRNA Details
Gene Symbol: 1700121M21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018973.1 | - | 1 | 118 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707326.1 | - | 1 | 118 | Chr. 15 - 99919874 - 99920299 [+] on Build GRCm38 |
