siRNA Details
Gene Symbol: 4930433E05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019593.1 | - | 2 | 181 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV263001.1 | - | 2 | 140 | Chr. 14 - 92861254 - 92866525 [-] on Build GRCm38 |
