siRNA Details
Gene Symbol: 4930434O05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019595.1 | - | 1 | 295 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714871.1 | - | 1 | 292 | Chr. 6 - 142989612 - 142990002 [+] on Build GRCm38 |
