siRNA Details
Gene Symbol: 9130015L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040499.1 | 3, 4 | 273 | |||
GenBank mRNA | AK018627.1 | - | 3, 4 | 274 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720808.1 | - | 3, 4 | 274 | Chr. 2 - 160016653 - 160040620 [+] on Build GRCm38 |
