siRNA Details
Gene Symbol: 4930452A19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045433.1 | 4 | 491 | |||
GenBank mRNA | AK019626.1 | - | 4 | 492 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714991.1 | - | 4 | 492 | Chr. 17 - 9750751 - 9775866 [-] on Build GRCm38 |
