siRNA Details
Gene Symbol: Gm36693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052522.1 | - | 4 | 429 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB660349.1 | - | 4 | 429 | Chr. 19 - 47199189 - 47202032 [-] on Build GRCm38 |
