siRNA Details
Gene Symbol: 5730564C23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019973.1 | - | 1 | 312 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717643.1 | - | 1 | 305 | Chr. 8 - 26051626 - 26052539 [-] on Build GRCm38 |
