siRNA Details
Gene Symbol: 4933405D12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016661.1 | - | 685 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716349.1 | - | 5, 6 | 685 | Chr. 3 - 122865158 - 122924116 [-] on Build GRCm38 |
