siRNA Details
Gene Symbol: Gm6297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077221.1 | 3 | 1179 | |||
NR_077226.1 | 3 | 1341 | ||||
GenBank mRNA | AK139286.1 | - | 1 | 2430 | ||
AK139295.1 | - | 2 | 1866 | |||
AK172127.1 | - | 1 | 1977 | |||
BB260315.2 | - | 1 | 209 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB810164.1 | - | 1 | 38 | Chr. 4 - 40720059 - 40720474 [-] on Build GRCm38 |
