siRNA Details
Gene Symbol: Gm6297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077221.1 | 3 | 240 | |||
NR_077226.1 | 3 | 402 | ||||
GenBank mRNA | AK139286.1 | - | 1 | 1491 | ||
AK139295.1 | - | 2 | 926 | |||
AK172127.1 | - | 1 | 1038 | |||
BE656271.1 | - | 3 | 402 | |||
CX215998.1 | - | 3 | 240 |
