siRNA Details
Gene Symbol: D530034E23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021308.1 | - | 1 | 431 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY730656.1 | - | 1 | 431 | Chr. 4 - 33402104 - 33402718 [+] on Build GRCm38 |
