siRNA Details
Gene Symbol: D530034E23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021308.1 | - | 1 | 266 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DV044340.1 | - | 1 | 436 | Chr. 4 - 33401950 - 33402732 [+] on Build GRCm38 |
