siRNA Details
Gene Symbol: 5430434N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017393.1 | - | 1 | 491 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717207.1 | - | 1 | 487 | Chr. 16 - 42510228 - 42510875 [-] on Build GRCm38 |
