siRNA Details
Gene Symbol: 1700071M16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045444.1 | 1 | 174 | |||
GenBank mRNA | AK006933.1 | - | 370 | |||
AK133239.1 | - | 1 | 174 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706947.1 | - | 2 | 370 | Chr. 17 - 43591160 - 43592432 [-] on Build GRCm38 |
