siRNA Details
Gene Symbol: 1700012I11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045140.1 | 1, 2 | 317 | |||
GenBank mRNA | AK005914.1 | - | 1, 2 | 318 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706053.1 | - | 321 | Chr. - - [-] on Build |
