siRNA Details
Gene Symbol: 1700012I11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045140.1 | 6 | 621 | |||
GenBank mRNA | AK005914.1 | - | 6 | 622 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706053.1 | - | 625 | Chr. - - [-] on Build |
