siRNA Details
Gene Symbol: 6230414M07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018087.1 | - | 1 | 304 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY718207.1 | - | 1 | 304 | Chr. 11 - 16958789 - 16959650 [+] on Build GRCm38 |
