siRNA Details
Gene Symbol: 9430078K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020496.1 | - | 3 | 403 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721865.1 | - | 3 | 403 | Chr. 12 - 79917457 - 79924733 [-] on Build GRCm38 |
