siRNA Details
Gene Symbol: B830008H07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046787.1 | - | 1 | 2040 | ||
AK135629.1 | - | 1 | 1360 | |||
BB333194.2 | - | 1 | 161 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV356187.2 | - | 1 | 115 | Chr. 7 - 45470991 - 45471617 [-] on Build GRCm38 |
