siRNA Details
Gene Symbol: 9230102O04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040511.1 | 4 | 718 | |||
GenBank mRNA | AK020309.1 | - | 4 | 719 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR517867.1 | - | 2 | 213 | Chr. 2 - 9883019 - 9883923 [-] on Build GRCm38 |
