siRNA Details
Gene Symbol: 9230102O04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040511.1 | 4 | 637 | |||
GenBank mRNA | AK020309.1 | - | 4 | 638 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DV047866.1 | - | 4 | 711 | Chr. 2 - 9883042 - 9889594 [-] on Build GRCm38 |
