siRNA Details
Gene Symbol: 6720482G16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020168.1 | - | 1 | 247 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719454.1 | - | 1 | 247 | Chr. 3 - 58526930 - 58527602 [+] on Build GRCm38 |
