siRNA Details
Gene Symbol: Gm2694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082312.1 | - | 2 | 407 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB650056.1 | - | 2 | 407 | Chr. 8 - 87473157 - 87503343 [+] on Build GRCm38 |
