siRNA Details
Gene Symbol: 1810035K13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019027.1 | - | 2 | 331 | ||
AV056323.1 | - | 1 | 276 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707721.1 | - | 2 | 331 | Chr. 19 - 58695521 - 58696181 [+] on Build GRCm38 |
