siRNA Details
Gene Symbol: Gm16725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040746.1 | - | 2 | 424 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB636371.1 | - | 2 | 420 | Chr. 8 - 18676381 - 18678054 [-] on Build GRCm38 |
